missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASP Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99085B
This item is not returnable.
View return policy
Description
WASP Polyclonal antibody specifically detects WASP in Human samples. It is validated for Western Blot
Specifications
| WASP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| IMD2THC, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked) | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human WASP. | |
| 0.1 mL | |
| Signal Transduction | |
| 7454 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction