Learn More
Miele Professional™ Demineralisation Cartridge LP 2800
Product Code. 15427724 Shop All Miele Products
Click to view available options
:
through flow volume: 800 l/h; model: resins
Product Code. 15427724
Brand: Miele Professional™ 9782590
Only null left
Add to basket
Add to basket
Demineralisation cartridge LP 2800, empty, for the manual filling of single use resins
  • Max. through flow volume: 800 l/h
  • Max water inlet temperature 40 °C - pressure from 0.5-10 bar
  • Resin content 19 litres
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel, Plastic
Product Type Water Pre-treatment
Volume (Metric) 19 L
Color Stainless Steel, Black
For Use With (Equipment) Washer-disinfectors
Diameter (Metric) 230 mm
Height (Metric) 570 mm