missing translation for 'onlineSavingsMsg'
Learn More

Aalborg™ Water Direct Glass Flow Tube

Product Code. 15264023
Change view
Click to view available options
Waterflow Rate:
1.2 L/min.
10 gal./hr.
100 mL/min.
2 L/min.
20 mL/min.
200 mL/min.
250 mL/min.
50 mL/min.
500 mL/min.
Width (Metric):
65 mm
150 mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Waterflow Rate Width (Metric) unitSize
15264023 10 gal./hr. 65 mm Each
11778258 1.2 L/min. 65 mm Each
11768258 500 mL/min. 65 mm Each
11783199 2 L/min. 150 mm Each
11758258 250 mL/min. 65 mm Each
11788338 1.2 L/min. 150 mm Each
11708338 50 mL/min. 150 mm Each
11793199 200 mL/min. 150 mm Each
11718338 100 mL/min. 150 mm Each
11703209 20 mL/min. 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 15264023 Supplier Aalborg™ Supplier No. FT06412GLVN
  • ONLINE EXCLUSIVE PRICE
    € 93.45
    € 57.12 / Pack of 1
    Save € 36.33
    38% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Aalborg™ Water Direct Glass Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.

  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.

Specifica

Type Water Direct Flow Tube
Width (Metric) 65 mm
Waterflow Rate 10 gal./hr.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.