Learn More
Hach Lange™ Water Hardness Cuvette Test
Product Code. 10743615
Click to view available options
No. of Tests:
25
Unit Size:
25 tests
Product Code. 10743615
Brand: Hach Lange™ LCK327
Only null left
Add to basket
Add to basket
Order Info
ShelfLife
Specifications
No. of Tests 25
Storage Requirements 15°C to 25°C
Test Method Metalphthalein
Type Water Hardness Cuvette Test