Learn More
Hach Lange™ Water Hardness Cuvette Test
Click to view available options
No. of Tests:
25
Unit Size:
25 tests
Description
Order Info
ShelfLife
Specifications
Specifications
| No. of Tests | 25 |
| Storage Requirements | 15°C to 25°C |
| Test Method | Metalphthalein |
| Type | Water Hardness Cuvette Test |