Learn More
LABPLAS Water Sampling Kit Writing
Product Code. p-200102234
Click to view available options
Volume (Metric):
100 mL
300 mL
500 mL
1000 mL
Unit Size:
Pack of 100
Product Code. 17910245
Brand: LABPLAS KWS21100
Only null left
Add to basket
Add to basket
Sterile water sampling bag with writing area for easy labeling.
  • Water Sampling Kit
  • Includes sodium thiosulfate tablet
  • Clear, with writing area for easy labeling
Specifications
Color Clear with White Block
Volume (Metric) 100 mL
Includes 10 mg Sodium Thiosulfate Tablet