Learn More
Kolmi Hopen™ Waterproof CPE Shoecovers
Wear these CPE shoecovers in the industrial or catering settings.
Brand: Kolmi Hopen™ H10150
Additional Details : Weight : 8.17000kg
Description
- Disposable and waterproof
- Intended to be worn by visitors, patients or caregivers
- Can be used as an object of environmental control in HACCP approaches and various industries
- H10150 model is thicker and more resistant; can be used in static work situations