Learn More
Watson-Marlow Manifold Tubing - PVC
Product Code. 10510176
Click to view available options
Diameter (Metric) Inner:
0.13 mm
0.19 mm
0.25 mm
0.38 mm
0.5 mm
0.63 mm
0.76 mm
1.02 mm
1.14 mm
1.29 mm
1.42 mm
1.52 mm
1.65 mm
1.85 mm
2.54 mm
2.79 mm
Color:
Black/Black
Blue/Blue
Gray/Gray
Green/Green
Orange/Black
Orange/Blue
Orange/Green
Orange/Red
Orange/White
Orange/Yellow
Purple/Orange
Purple/White
Red/Red
White
Yellow
Yellow/Blue
Unit Size:
Each
Pack of 12
Product Code. 10510176
Brand: Watson-Marlow 981.0063.000
Only null left
Add to basket
Add to basket
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner 1/50 in.
Diameter (Metric) Inner 0.63 mm
Material PVC
Thickness (English) 0.03 in.
Thickness (Metric) 0.8 mm
For Use With 323S/MC peristaltic pump
Tubing Size 0.63mm
Flow Rate 0.04 to 3.1 (four roller pump), 0.04 to 2.6 (eight roller pump)mL/min.
Color Orange/White
Product Type Pump Tubing