missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WAY 252623

Description
WAY 252623 is a potent LXR agonist (IC50 values are 24 and 179 nM for LXRβ and LXRα, respectively). Reduces total serum cholesterol and LDL cholesterol, and inhibits lesion growth in models of atherosclerosis. Increases ABCA1 and ABCG1 mRNA levels in rat peripheral blood cells. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 2-[(2-Chloro-4-fluorophenyl)methyl]-3-(4-fluorophenyl)-7-(trifluoromethyl)-2H-indazole |
| CAS | 875787-07-8 |
| Quantity | 10 mg |
| Target | LXR-like Receptor Agonists |
| Molecular Formula | C21H12ClF5N2 |
| Formula Weight | 422.78 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?