Learn More
WBP1 Polyclonal Antibody
GREENER_CHOICE
Product Code. 16557111
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16557111 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16557111 Supplier Invitrogen Supplier No. PA589083
WBP1 Polyclonal Antibody
WBP1 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen WBP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene WBP1
Gene Accession No. P97764, Q96G27
Gene Alias AI255152; WBP1; WBP-1; WW domain binding protein 1; WW domain-binding protein 1
Gene Symbols WBP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 170-269 of human WBP1 (NP_0366091).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22377, 23559, 297381
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG