missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP11 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97493FR
This item is not returnable.
View return policy
Description
WBP11 Polyclonal specifically detects WBP11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| WBP11 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| DKFZp779M1063, Npw38-binding protein, Npw38-binding protein NpwBP, NpwBP, NPWBPsplicing factor, PQBP1 and PP1 interacting, SH3 domain-binding protein SNP70, SIPP1WW domain-binding protein 11, SNP70, Splicing factor that interacts with PQBP-1 and PP1, WBP-11, WW domain binding protein 11 | |
| Produced in rabbits immunized with E. coli-derived Human WBP11 fragment. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51729 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction