missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WBP11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17149033
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17149033 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17149033 Supplier Invitrogen™ Supplier No. PA5111113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DIYKELRKLE VEYEQKRAQL SQYFDAVKNA QHVEVESIPL PDMPHAPSNI LIQDIPLPGA QPPSILKKTS AYGPPTRAVS ILPLLGHGVP RLP Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 100%.

Posttranslational modification of proteins by the addition of the small protein SUMO.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WBP11
Gene Accession No. Q9Y2W2
Gene Alias 2510026P17Rik; D6Wsu113e; DKFZp779M1063; hypothetical protein LOC402950; Npw38-binding protein; Npw38-binding protein NpwBP; NpwBP; PPP1R165; protein phosphatase 1, regulatory subunit 165; SH3 domain-binding protein; SH3 domain-binding protein SNP70; SIPP1; SNP70; Splicing factor that interacts with PQBP-1 and PP1; splicing factor, PQBP1 and PP1 interacting; Wbp11; WBP-11; WW domain binding protein 11; WW domain-binding protein 11; zgc:77390
Gene Symbols WBP11
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human WBP11. Recombinant protein control fragment (Product #RP-104707).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.