missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WBP2NL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561768
This item is not returnable.
View return policy
Description
Immunogen sequence: QSHTENRRGA LIPNGESLLK RSPNVELSFP QRSEGSNVFS GRKTGTLFLT SYRVIFITSC SISDPMLSFM MPFDLMTNLT VEQPVFAANF IKGTIQAAPY GGWEGQATFK LVFRNGDAIE FAQLMVKAA Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
May play a role in meotic resumption and pronuclear formation, mediated by a WW domain-signaling pathway during fertilization.
Specifications
| WBP2NL | |
| Polyclonal | |
| Unconjugated | |
| WBP2NL | |
| 4930521I23Rik; GRAMD7; PAWP; Postacrosomal sheath WW domain-binding protein; WBP2 N-terminal like; Wbp2nl; WW domain-binding protein 2-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 164684 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ICG8 | |
| WBP2NL | |
| Recombinant protein corresponding to Human WBP2NL. Recombinant protein control fragment (Product #RP-91524). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction