missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WBP5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15876843
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15876843 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15876843 Supplier Invitrogen™ Supplier No. PA568432

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WBP5 Polyclonal Antibody for IHC (P)

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TCEAL9
Gene Accession No. Q9UHQ7
Gene Alias pp21 homolog; TCEAL9; TCEA-like protein 9; transcription elongation factor A like 9; transcription elongation factor A protein-like 9; Transcription elongation factor S-II protein-like 9; WBP5; WBP-5; WEX6; WW domain binding protein 1; WW domain binding protein 5; WW domain-binding protein 5
Gene Symbols TCEAL9
Host Species Rabbit
Immunogen Full length Fusion protein of human WBP5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51186
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.