Learn More
WBSCR17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21280-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| WBSCR17 | |
| Polyclonal | |
| Unconjugated | |
| GALNT17 | |
| GalNAc T like protein 3, GALNT16, GALNT20, GALNTL3, pp GaNTase like protein 3, WBSCR17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 212996, 288611, 64409 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6IS24, Q7TT15 | |
| Galnt17 | |
| WBSCR17 Fusion Protein Ag15792 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |