Learn More
WBSCR17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21280-1-AP-150UL
 View more versions of this product
Product Code. 16800895
  • € 410.00 / 150µL
Specifications
WBSCR17
Polyclonal
Unconjugated
GALNT17
GalNAc T like protein 3, GALNT16, GALNT20, GALNTL3, pp GaNTase like protein 3, WBSCR17
Rabbit
Antigen Affinity Chromatography
RUO
212996, 288611, 64409
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6IS24, Q7TT15
Galnt17
WBSCR17 Fusion Protein Ag15792
150 μL
Primary
Human, Mouse, Rat