missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ WDFY3 Antibody (4G9H4), Novus Biologicals™
Mouse Monoclonal Antibody
€ 287.00 - € 511.00
Specifications
| Antigen | WDFY3 |
|---|---|
| Clone | 4G9H4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029491
|
Novus Biologicals™
NBP3-27153-0.025ml |
0.025 mL |
€ 287.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028562
|
Novus Biologicals™
NBP3-27153-0.1ml |
0.1 mL |
€ 511.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WDFY3 Monoclonal antibody specifically detects WDFY3 in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| WDFY3 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Autophagy, Cancer | |
| PBS | |
| 23001 | |
| IgG1 | |
| Protein G purified |
| 4G9H4 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ALFY, Autophagy-linked FYVE protein, KIAA0993MGC16461, WD repeat and FYVE domain containing 3, WD repeat and FYVE domain-containing protein 3, ZFYVE25 | |
| Purified recombinant fragment of human WDFY3 (AA: 3277-3526) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title