missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDHD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 425.25 - € 612.15
Specifications
| Antigen | WDHD1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692745
|
Novus Biologicals
NBP3-21217-100ul |
100 μg |
€ 648.00 € 612.15 / 100µL Save € 35.85 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669873
|
Novus Biologicals
NBP3-21217-25ul |
25 μg |
€ 450.00 € 425.25 / 25µL Save € 24.75 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WDHD1 Polyclonal antibody specifically detects WDHD1 in Human samples. It is validated for Western BlotSpecifications
| WDHD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 11169 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Acidic nucleoplasmic DNA-binding protein 1, AND1, AND-1, CHTF4, CTF4, CTF4, chromosome transmission fidelity factor 4 homolog, WD repeat and HMG-box DNA binding protein 1, WD repeat and HMG-box DNA-binding protein 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QTLNIVTWSPCGQYLAAGSINGLIIVWNVETKDCMERVKHEKGYAICGLAWHPTCGRISYTDAEGNLGLLENVCDPSGKTSSSKVSSRVEKDYNDLFDGDDMSNAGDFLNDNAVE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title