missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDHD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15789470 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15789470 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15789470 Supplier Invitrogen™ Supplier No. PA551517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QTLNIVTWSP CGQYLAAGSI NGLIIVWNVE TKDCMERVKH EKGYAICGLA WHPTCGRISY TDAEGNLGLL ENVCDPSGKT SSSKVSSRVE KDYNDLFDGD DMSNAGDFLN DNAVE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.

The protein encoded by this gene contains multiple N-terminal WD40 domains and a C-terminal high mobility group (HMG) box. WD40 domains are found in a variety of eukaryotic proteins and may function as adaptor/regulatory modules in signal transduction, pre-mRNA processing and cytoskeleton assembly. HMG boxes are found in many eukaryotic proteins involved in chromatin assembly, transcription and replication. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDHD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Wdhd1
Gene Accession No. O75717, P59328
Gene Alias A630092E18; Acidic nucleoplasmic DNA-binding protein 1; AND1; And-1; CHTF4; crm1; CRM1/XPO1 protein; CTF4; CTF4, chromosome transmission fidelity factor 4 homolog; D630024B06Rik; exportin 1 (CRM1 homolog); exportin 1 S homeolog; exportin-1; WD repeat and HMG-box DNA binding protein 1; WD repeat and HMG-box DNA-binding protein 1; WD repeat and HMG-box DNA-binding protein 1-like protein; WDHD1; XELAEV_18028477mg; Xpo; xpo1; xpo1.S
Gene Symbols Wdhd1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human WDHD1 (Product #RP-88907)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11169, 218973, 305827
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.