missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDPCP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA524271

Product Code. 13202149

  • € 578.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

WDPCP Polyclonal Antibody for Western Blot, IHC (P)

The C2ORF86 gene encodes an ortholog of Drosophila 'fritz,' a cytoplasmic WD40 repeat protein that controls planar cell polarity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WDPCP
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
O95876
WDPCP
KLH conjugated synthetic peptide between 676-705 amino acids from the C-terminal region of human CB086.
400 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
WDPCP
AV249152; Bardet-Biedl syndrome 15 protein; BBS15; C2orf86; CHDTHP; FRITZ; FRTZ; hFRTZ; homoloc-13; homolog-13; mFrtz; WD repeat containing planar cell polarity effector; WD repeat-containing and planar cell polarity effector protein; WD repeat-containing and planar cell polarity effector protein fritz homolog; WD repeat-containing protein C2orf86; WD repeat-containing protein C2orf86 homolog; Wdpcp
Rabbit
Antigen affinity chromatography, Protein A
RUO
51057
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.