missing translation for 'onlineSavingsMsg'
Learn More

WDR11 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93248-0.1ml

 View more versions of this product

Product Code. 18625940

  • € 470.00 / 0.01mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

WDR11 Polyclonal antibody specifically detects WDR11 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WDR11
Polyclonal
Western Blot 1:1000-1:2000
Bromodomain and WD repeat-containing protein 2, BRWD2DKFZp434L1715, DR11, FLJ10506, FLJ42531, KIAA1351bromodomain and WD repeat domain containing 2, WD repeat domain 11, WD repeat domain 15, WD repeat-containing protein 11, WD repeat-containing protein 15, WDR15
Recombinant fusion protein containing a sequence corresponding to amino acids 975-1224 of human WDR11 (NP_060587.8). YFQKFQLERVNLQEVKRSTYDHTRKCTDQLLLLGQTDRAVQLLLETSADNQHYYCDSLKACLVTTVTSSGPSQSTIKLVATNMIANGKLAEGVQLLCLIDKAADACRYLQTYGEWNRAAWLAKVRLNPEECADVLRRWVDHLCSPQVNQKSKALLVLLSLGCFFSVAETLHSMRYFDRAALFVEACLKYGAFEVTEDTEKLITAIYADYARSLKNLGFKQGAVLFASKAGAAGKDLLNELESPKEEPIEE
0.1 mL
Cancer
55717
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.