missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR13, Mouse anti-Human, Clone: 1G9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00064743-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: GTLQLKRSFPIEQSSHPVRSIFCPLMSFRQGACVVTGSEDMCVHFFDVERAAKAAVNKLQGHSAPVLDVSFNCDESLLASSDASGMVIVWRREQSpecifications
| WDR13 | |
| Monoclonal | |
| Unconjugated | |
| WD repeat domain 13 | |
| DKFZp779C2057/FLJ20563/MG21 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1G9 | |
| In 1x PBS, pH 7.4 | |
| NM_017883 | |
| WDR13 | |
| WDR13 (NP_060353.2, 391 a.a. ∼ 484 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 64743 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction