missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR18 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81877DL594
This item is not returnable.
View return policy
Description
WDR18 Polyclonal antibody specifically detects WDR18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| WDR18 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| MGC2436, R32184_1, WD repeat domain 18, WD repeat-containing protein 18 | |
| WDR18 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human WDR18. The immunogen is located within the last 50 amino acids of WDR18. Amino Acid Squence: VRNLRKINRDLFDFSTR | |
| 0.1 mL | |
| Cell Biology | |
| 57418 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction