missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR19 Polyclonal Antibody

Product Code. 16381345
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16381345 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16381345 Supplier Invitrogen™ Supplier No. PA584395

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HMEVEFAIRV YRRIGNVGIV MSLEQIKGIE DYNLLAGHLA MFTNDYNLAQ DLYLASSCPI AALEMRRDLQ HWDSALQLAK HLAPDQ Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains six WD repeats, a clathrin heavy-chain repeat, and three transmembrane domains. This gene is conserved from C. elegans to human. It may participate in androgen-regulated signaling mechanisms or in the vesicular trafficking of androgen-regulated secretory processes. Alternatively spliced transcript variants encoding distinct isoforms have been reported but the full-length nature of one of these variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR19
Gene Accession No. Q8NEZ3
Gene Alias ATD5; C330027H04Rik; CED4; D330023L08Rik; DYF2; DYF-2; Ift144; Intraflagellar transport 144 homolog; Kiaa1638; mKIAA1638; NPHP13; ORF26; Oseg6; PWDMP; RGD1306997; SRTD5; WD repeat domain 19; WD repeat membrane protein PWDMP; WD repeat-containing protein 19; WDR19
Gene Symbols WDR19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR19. Recombinant protein control fragment (Product #RP-105116).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57728
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.