missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR20 Antibody (1D10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00091833-M01
This item is not returnable.
View return policy
Description
WDR20 Monoclonal antibody specifically detects WDR20 in Human samples. It is validated for Western Blot, ELISA
Specifications
| WDR20 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DMR, FLJ33659, MGC33177, MGC33183, Protein DMR, WD repeat domain 20, WD repeat-containing protein 20 | |
| WDR20 (AAH28387, 1 a.a. ∽ 569 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MATEGGGKEMNEIKTQFTTREGLYKLLPHSEYSRPNRVPFNSQGSNPVRVSFVNLNDQSGNGDRLCFNVGRELYFYIYKGVRKAADLSKPIDKRIYKGTQPTCHDFNHLTATAESVSLLVGFSAGQVQLIDPIKKETSKLFNEERLIDKSRVTCVKWVHGSESLFLVAHSSGNMYLYNVEHTCGTTAPHYQLLKQGESFAVHTCKSKSTRNPLLKWTVGEGALNEFAFSPDGKFLACVSQDGFLRVFNFDSVELHGTMKSYFGGLLCVCWSPDGKYIVTGGEDDLVTVWSFVDCRVIARGHGHKSWVSVVAFDPYTTSVEEGDPMEFSGSDEDFQDLLHFGRDRANSTQSRLSKRNSTDSRPVSVTYRFGSVGQDTQLCLWDLTEDILFPHQPLSRARTHTNVMNATSPPAGSNGNSVTTPGNSVPPPLPRSNSLPHSAVSNAGSKSSVMDGAIASGVSKFATLSLHDRKERHHEKDHKRNHSMGHISSKSSDKLNLVTKTKTDPAKTLGTPLCPRMEDVPLLEPLICKKIAHERLTVLIFLEDCIVTACQEGFICTWGRPGKVVSFNP | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1D10 | |
| Western Blot 1:500, ELISA | |
| AAH28387 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 91833 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction