missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR23 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16537871
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16537871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16537871 Supplier Invitrogen™ Supplier No. PA590526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: LO2, HeLa, Mouse brain, Mouse heart, Rat brain.

WDR23 contains 7 WD repeats. The function of WDR23 remains unknown.This gene encodes a WD repeat-containing protein. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR23
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Dcaf11
Gene Accession No. Q5M9G8, Q8TEB1, Q91VU6
Gene Alias 0710008A13Rik; C76035; D14Ucla1; Dacf11; DCAF11; DDB1 and CUL4 associated factor 11; DDB1- and CUL4-associated factor 11; DKFZp779A1629; GL014; GLO14; PRO2389; WD repeat domain 23; WD repeat domain 23 isoform 1; WD repeat-containing protein 23; WDR23; WDR23 protein
Gene Symbols Dcaf11
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28199, 305895, 80344
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.