missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR26 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26629C
This item is not returnable.
View return policy
Description
WDR26 Polyclonal antibody specifically detects WDR26 in Human samples. It is validated for Western Blot
Specifications
| WDR26 | |
| Polyclonal | |
| Western Blot | |
| MIP2, myocardial ischemic preconditioning up-regulated protein 2, WD repeat domain 26, WD repeat-containing protein 26 | |
| The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human WD repeat domain 26 using the numbering given in entry BAG10986.1 (GeneID 80232). | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 80232 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction