Learn More
Invitrogen™ WDR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140035
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ARKKAKLHSS KGEEEDPEVN VEMSLQDEIQ RVTNIKTSAK IKSFDLIHSP HGELKAVFLL QNNLVELYSL NPSLPTPQPV RTSRITIG Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 83%.
This gene encodes a nuclear protein containing 10 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, which usually include a trp-asp at the C-terminal end. Proteins belonging to the WD repeat family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. [provided by RefSeq, Jul 2008].
Specifications
| WDR3 | |
| Polyclonal | |
| Unconjugated | |
| WDR3 | |
| DIP2; dJ776P7.2 (WD repeat domain 3); UTP12; WD repeat domain 3; WD repeat-containing protein 3; WDR3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10885 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UNX4 | |
| WDR3 | |
| Recombinant protein corresponding to Human WDR3. Recombinant protein control fragment (Product #RP-109309). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.