missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR35 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-82058IR
This item is not returnable.
View return policy
Description
WDR35 Polyclonal antibody specifically detects WDR35 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| WDR35 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| CED2, IFT121, WD repeat domain 35 | |
| WDR35 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human WDR35. The immunogen is located within amino acids 60 - 110 of WDR35. Amino Acid Squence: TWNEQYQKLTTSDENG | |
| 0.1 mL | |
| Cell Biology | |
| 57539 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction