missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557924
This item is not returnable.
View return policy
Description
Immunogen sequence: FRDPSIHSVN VFQGHTDTVT SAVFTVGDNV VSGSDDRTVK VWDLKNMRSP IATIRTDSAI NRINVCVGQK IIALPHDNRQ VRLFDMSGVR LARLPRSSRQ GHRRMVCCS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
WDR37 encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes.
Specifications
| WDR37 | |
| Polyclonal | |
| Unconjugated | |
| WDR37 | |
| fj52e09; im:7144367; Kiaa0982; RP11-529L18.2; WD repeat domain 37; WD repeat-containing protein 37; Wdr37; wu:fj52e09 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 207615, 22884, 307075 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8CBE3, Q9Y2I8 | |
| WDR37 | |
| Recombinant protein corresponding to Human WDR37. Recombinant protein control fragment (Product #RP-97003). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction