missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR44 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-68250
This item is not returnable.
View return policy
Description
WDR44 Polyclonal specifically detects WDR44 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| WDR44 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| DKFZp686L20145, FLJ13230, MGC26781, RAB11BP, rabphilin-11, RPH11, WD repeat domain 44, WD repeat-containing protein 44 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54521 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q5JSH3 | |
| WDR44 | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human WD repeat domain 44 using the numbering given in entry NP_061918.3 (GeneID 54521). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction