missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR44 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15998194
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15998194 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15998194 Supplier Invitrogen™ Supplier No. PA565454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VIGTYDGRCIF YDTEHLKYHT QIHVRSTRGR NKVGRKITGI EPLPGENKIL VTSNDSRIRL YDLRDLSLSM KYKG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR44
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR44
Gene Accession No. Q5JSH3
Gene Alias RAB11BP; rabphilin-11; RPH11; WD repeat domain 44; WD repeat-containing protein 44; WDR44
Gene Symbols WDR44
Host Species Rabbit
Immunogen Recombinant Human WDR44. Recombinant protein control fragment (Product #RP-106133).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54521
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.