missing translation for 'onlineSavingsMsg'
Learn More

WDR45 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18575325 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18575325 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18575325 Supplier Novus Biologicals Supplier No. NBP318255AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WDR45 Polyclonal antibody specifically detects WDR45 in Human, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR45
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias JM5, WD repeat domain 45, WD repeat domain phosphoinositide-interacting protein 4, WD repeat-containing protein 45, WD45 repeat protein interacting with phosphoinositides 4, WDRX1, WDRXI4, WIPI-4, X-linked 1
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminal of Human WDR45(aa. 312-322)
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 11152
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.