missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR5, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31340.50ug
This item is not returnable.
View return policy
Specifications
| WDR5 | |
| Polyclonal | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| WD repeat domain 5 | |
| WDR5 | |
| Recombinant protein corresponding to human WDR5. | |
| RUO | |
| 11091 | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| In PBS (0.05% sodium azide, 0.05% proclin 300). | |
| BIG-3/SWD3 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction