missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR54 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97811AF750
This item is not returnable.
View return policy
Description
WDR54 Polyclonal antibody specifically detects WDR54 in Human samples. It is validated for Western Blot
Specifications
| WDR54 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84058 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| FLJ12953, WD repeat domain 54, WD repeat-containing protein 54 | |
| Produced in rabbits immunized with E. coli-derived Human WDR54 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction