missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR61 Monoclonal Antibody (OTI4C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807587S
This item is not returnable.
View return policy
Description
WDR61 Monoclonal antibody specifically detects WDR61 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| WDR61 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P61371 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3670 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C4 | |
| Unconjugated | |
| ISL1 | |
| 2700038L12Rik; 2810418I05Rik; meiotic recombination protein REC14; meiotic recombination REC14 protein homolog; REC14; recombination protein REC14; RGD1308228; SKI8; SKI8 homolog; WD repeat domain 61; WD repeat-containing protein 61; WD repeat-containing protein 61, N-terminally processed; WDR61 | |
| Human recombinant protein fragment corresponding to amino acids 134-349 of human ISL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction