missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR61 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118359
This item is not returnable.
View return policy
Description
WDR61 Polyclonal Antibody for Western Blot, ICC/IF, IP
SKI8 is a component of the PAF and SKI complex which is composed of SKI2, SKI3 and SKI8. These are involved in transcriptional regulation and events downstream of RNA synthesis. Interaction of the SKI complex with SKI7 links the SKI complex to the exosome in order to perform mRNA degradation.
Specifications
| WDR61 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9GZS3 | |
| WDR61 | |
| E. coli-derived Human WDR61 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| WDR61 | |
| 2700038L12Rik; 2810418I05Rik; meiotic recombination protein REC14; meiotic recombination REC14 protein homolog; REC14; recombination protein REC14; RGD1308228; SKI8; SKI8 homolog; WD repeat domain 61; WD repeat-containing protein 61; WD repeat-containing protein 61, N-terminally processed; WDR61 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 80349 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction