Learn More
Invitrogen™ WDR7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144042
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Daudi whole cell, human MOLT-4 whole cell, human HEL whole cell, rat brain tissue, mouse brain tissue. IHC: mouse brain tissue, rat brain tissue. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD) that may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. The encoded protein forms the beta subunit of rabconnectin-3 and binds directly with Rab3A GDP/GTP exchange protein and indirectly with Rab3A GDP/GTP activating protein; these proteins are regulators of Rab3 small G protein family members involved in control of the calcium-dependent exocytosis of neurotransmitters. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| WDR7 | |
| Polyclonal | |
| Unconjugated | |
| WDR7 | |
| AI462727; Kiaa0541; LOW QUALITY PROTEIN: WD repeat-containing protein 7; mKIAA0541; rabconnectin-3 beta; rabconnectin-3beta; TGF-beta resistance associated; TGF-beta resistance-associated protein TRAG; TRAG; WD repeat domain 7; WD repeat-containing protein 7; WD repeat-containing protein 7-like protein; Wd7; WDR7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 104082, 23335, 66031 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q920I9, Q9ERH3, Q9Y4E6 | |
| WDR7 | |
| E.coli-derived human WDR7 recombinant protein (Position: Q258-Q1292). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.