missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR74 Antibody (OTI4B2), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74867JF646
This item is not returnable.
View return policy
Description
WDR74 Monoclonal specifically detects WDR74 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| WDR74 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| WDR74 | |
| Recombinant protein fragment corresponding to amino acids 114-385 of human WDR74 (NP_060563) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4B2 | |
| Western Blot, Immunohistochemistry | |
| FLJ10439, FLJ21730, NOP seven-associated protein 1, NSA1, WD repeat domain 74, WD repeat-containing protein 74 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 54663.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction