missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR74 Monoclonal Antibody (OTI3D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800502S
This item is not returnable.
View return policy
Description
WDR74 Monoclonal antibody specifically detects WDR74 in Canine, Human samples. It is validated for Western BlotSpecifications
| WDR74 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50897 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 475316, 5538 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3D10 | |
| Unconjugated | |
| Ppt1 | |
| 5730436H21Rik; AA407588; NOP seven-associated protein 1; NSA1; WD repeat domain 74; WD repeat-containing protein 74; Wdr74 | |
| Human recombinant protein fragment corresponding to amino acids 100-306 of human PPT1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction