missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR74 Monoclonal Antibody (OTI3D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800502
This item is not returnable.
View return policy
Description
The yeast nucleolar protein NOP7 is necessary for the maturation of 66S preribosomes and interacts with numerous other proteins. One such protein is an essential, conserved WD repeat protein, NOP seven-associated protein 1 (NSA1), that is also required for the yeast 66S ribosome assembly. NSA1 is also associated with the AAA ATPase Rix7, and release of NSA1 from a novel late nucleolar pre-60S requires the Rix7 function. NSA1 has also been found upregulated in mammalian cancer cells, suggesting it may also play a role in cell proliferation.Specifications
| WDR74 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6RFH5 | |
| WDR74 | |
| Human recombit protein fragment corresponding to amino acids 114-385 of human WDR74 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3D10 | |
| Unconjugated | |
| WDR74 | |
| WD repeat domain 74 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54663 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction