Learn More
Invitrogen™ WDR74 Monoclonal Antibody (OTI4B2)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526448
Product Code. 15956044
  • € 588.00 / 100µL
Description
WDR74 Monoclonal Antibody for Western Blot, IHC (P)
The yeast nucleolar protein NOP7 is necessary for the maturation of 66S preribosomes and interacts with numerous other proteins. One such protein is an essential, conserved WD repeat protein, NOP seven-associated protein 1 (NSA1), that is also required for the yeast 66S ribosome assembly. NSA1 is also associated with the AAA ATPase Rix7, and release of NSA1 from a novel late nucleolar pre-60S requires the Rix7 function. NSA1 has also been found upregulated in mammalian cancer cells, suggesting it may also play a role in cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WDR74
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6RFH5, Q8VCG3
WDR74
Human recombinant protein fragment corresponding to amino acids 114-385 of WDR74 produced in E.coli.
100 μL
Primary
Human, Mouse, Rat, Canine, Monkey
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI4B2
Unconjugated
WDR74
5730436H21Rik; AA407588; NOP seven-associated protein 1; NSA1; WD repeat domain 74; WD repeat-containing protein 74; Wdr74
Mouse
Affinity chromatography
RUO
107071, 483778, 54663, 690229
-20°C, Avoid Freeze/Thaw Cycles