missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR77 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38391-20ul
This item is not returnable.
View return policy
Description
WDR77 Polyclonal antibody specifically detects WDR77 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| WDR77 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Androgen receptor cofactor p44, MEP-50, MEP50Nbla10071, methylosome protein 50, MGC2722, p44, p44/Mep50, RP11-552M11.3, WD repeat domain 77, WD repeat-containing protein 77 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 153-243 of human WDR77 (NP_077007.1).,, Sequence:, DLAQQVVLSSYRAHAAQVTCVAASPHKDSVFLSCSEDNRILLWDTRCPKPASQIGCSAPGYLPTSLAWHPQQSEVFVFGDENGTVSLVDTK | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 79084 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction