missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR77 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 539.00
Specifications
| Antigen | WDR77 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18487810
|
Novus Biologicals
NBP1-82778-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18299656
|
Novus Biologicals
NBP1-82778 |
0.1 mL |
€ 539.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WDR77 Polyclonal specifically detects WDR77 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| WDR77 | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 79084 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SEDCSIAVLDSSLSELFRSQAHRDFVRDATWSPLNHSLLTTVGWDHQVVHHVVPTEPLPAPGPASVTE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Androgen receptor cofactor p44, MEP-50, MEP50Nbla10071, methylosome protein 50, MGC2722, p44, p44/Mep50, RP11-552M11.3, WD repeat domain 77, WD repeat-containing protein 77 | |
| WDR77 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title