missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR77 Monoclonal Antibody (OTI5E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809719
This item is not returnable.
View return policy
Description
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles.Specifications
| WDR77 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQA1 | |
| WDR77 | |
| Human recombit protein fragment corresponding to amino acids 1-250 of human WDR77 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5E9 | |
| Unconjugated | |
| WDR77 | |
| HKMT1069, MEP-50, MEP50, Nbla10071, p44, p44/Mep50 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79084 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction