missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR77 Monoclonal Antibody (OTI5E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809719S
This item is not returnable.
View return policy
Description
WDR77 Monoclonal antibody specifically detects WDR77 in Human samples. It is validated for Western BlotSpecifications
| WDR77 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75368 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6451 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5E9 | |
| Unconjugated | |
| SH3BGRL | |
| 2610003I18Rik; 2610312E17Rik; Ac2-269; androgen receptor cofactor p44; C79984; HKMT1069; MEP50; MEP-50; Methylosome protein 50; MGC2722; Nbla10071; OTTHUMP00000013473; p44; p44/Mep50; RGD1310479; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat domain 77; WD repeat-containing protein 77; WD45; WDR77 | |
| Full length human recombinant protein of human SH3BGRL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction