missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MORG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526580
This item is not returnable.
View return policy
Description
MORG1 Polyclonal Antibody for Western Blot
MORG1 is a member of the WD-40 protein family. The protein is proposed to function as a molecular scaffold for various multimeric protein complexes. The protein associates with several components of the extracellular signal-regulated kinase (ERK) pathway, and promotes ERK activity in response to serum or other signals. The protein also interacts with egl nine homolog 3 (EGLN3, also known as PHD3) and regulates expression of hypoxia-inducible factor 1, and has been purified as part of the spliceosome.
Specifications
| MORG1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9BRX9 | |
| WDR83 | |
| KLH conjugated synthetic peptide between 126-154 amino acids from the central region of human MORG1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| WDR83 | |
| 1500041N16Rik; MAPK cascade scaffold protein; MAPK organizer 1; MGC4238; Mitogen-activated protein kinase organizer 1; MORG1; RGD1306947; WD repeat domain 83; WD repeat domain containing 83; WD repeat domain-containing protein 83; WD repeat protein Morg1; WDR83 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 84292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction