missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR88 Polyclonal Antibody

Product Code. 15948354
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15948354 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15948354 Supplier Invitrogen™ Supplier No. PA565611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ILSASKDRTMR LWNIEEIDEI PLVIKYKKAV GLKLKQCERC DRPFSIFKSD TSSEMFTQCV FCRIDTRGLP ADTS Highest antigen sequence identity to the following orthologs - mouse 24%, rat 55%.

WDR88 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR88
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR88
Gene Accession No. Q6ZMY6
Gene Alias PQQ repeat and WD repeat domain containing; PQQ repeat and WD repeat-containing protein; PQWD; WD repeat domain 88; WD repeat-containing protein 88; WDR88
Gene Symbols WDR88
Host Species Rabbit
Immunogen Recombinant Human WDR88. Recombinant protein control fragment (Product #RP-106277).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126248
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.