Learn More
Invitrogen™ WDR88 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598512
Product Code. 16591332
  • € 473.00 / 100µg
Description
WDR88 Polyclonal Antibody for Western Blot, IHC (P), ELISA
WDR88 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WDR88
Polyclonal
Unconjugated
WDR88
PQQ repeat and WD repeat domain containing; PQQ repeat and WD repeat-containing protein; PQWD; WD repeat domain 88; WD repeat-containing protein 88; WDR88
Rabbit
Protein G
RUO
126248
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q6ZMY6
WDR88
Recombinant Human WD repeat-containing protein 88 protein (1-250AA).