missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR90 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-31888-25ul
This item is not returnable.
View return policy
Description
WDR90 Polyclonal specifically detects WDR90 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| WDR90 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q96KV7 | |
| WDR90 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FKEFKSTATWLQFPLVLEARTPQRDLVGLAPSGARWTCLQLDLQDVLLVYLNRCYGHLKSIRLCASLLVRNLYTSDLCFEPAISGAQWAKLPVT | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C16orf15, C16orf16, C16orf17, C16orf18, C16orf19, chromosome 16 open reading frame 15, chromosome 16 open reading frame 17, chromosome 16 open reading frame 18, chromosome 16 open reading frame 19, FLJ36483, FLJ44660, KIAA1924chromosome 16 open reading frame 16, WD repeat domain 90, WD repeat-containing protein 90 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 197335 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction