missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR91 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-77307H
This item is not returnable.
View return policy
Description
WDR91 Polyclonal antibody specifically detects WDR91 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| WDR91 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| HSPC049, WD repeat domain 91, WD repeat-containing protein 91 | |
| The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human WD repeat domain 91 using the numbering given in entry NP_054868.2 (GeneID 29062). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 29062 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction