missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR92 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76282C
This item is not returnable.
View return policy
Description
WDR92 Polyclonal antibody specifically detects WDR92 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| WDR92 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| FLJ31741, LOC116143, Monad, WD repeat domain 92, WD repeat-containing protein 92, WD repeat-containing protein Monad | |
| Antibody was raised against a 21 amino acid synthetic peptide near the center of human WDR92. The immunogen is located within amino acids 140 - 190 of WDR92. Amino Acid Squence: CDPRQKDDPVANMEPVQGENKR | |
| 0.1 mL | |
| Apoptosis | |
| 116143 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction